Skip to Content
Merck

BDPH9BCDCF3E

Bide Pharmatech

2,4-Dichloro-5-cyclopropylpyrimidine

Synonym(s):

2,4-dichloro-5-cyclopropylpyrimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H6N2Cl2
CAS Number:
Molecular Weight:
189.04
MDL number:
Product BDPH9BCDCF3E is not currently sold in your country.


Inchi Code

1S/C7H6Cl2N2/c8-6-5(4-1-2-4)3-10-7(9)11-6/h3-4H,1-2H2

Inchi Key

KKSKKVUKDFJQMS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or Semi-solid or liquid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library