Skip to Content

BDPH9BCDD536

Bide Pharmatech

2,6-Dichloro-4-methylaniline

Synonym(s):

2,6-dichloro-4-methylaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7N1Cl2
CAS Number:
Molecular Weight:
176.04
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7Cl2N/c1-4-2-5(8)7(10)6(9)3-4/h2-3H,10H2,1H3

Inchi Key

HMUDNHJDRNNRIE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Pale-yellow to Yellow-brown Solid



Still not finding the right product?

Explore all of our products under 2,6-Dichloro-4-methylaniline


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library