Skip to Content
Merck

BDPH9BCDEB16

Bide Pharmatech

3-Fluoro-4-sulfanylbenzoic acid

Synonym(s):

3-fluoro-4-sulfanylbenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5O2S1F1
CAS Number:
Molecular Weight:
172.18
MDL number:
Product BDPH9BCDEB16 is not currently sold in your country.


Inchi Code

1S/C7H5FO2S/c8-5-3-4(7(9)10)1-2-6(5)11/h1-3,11H,(H,9,10)

Inchi Key

YROBFVBSJCRPDE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 3-Fluoro-4-sulfanylbenzoic acid




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library