Skip to Content
Merck

BDPH9BCDEEEA

Bide Pharmatech

2-Bromo-4,5-dimethylpyridine

Synonym(s):

2-bromo-4,5-dimethylpyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H8N1Br1
CAS Number:
Molecular Weight:
186.05
MDL number:
Product BDPH9BCDEEEA is not currently sold in your country.


Inchi Code

1S/C7H8BrN/c1-5-3-7(8)9-4-6(5)2/h3-4H,1-2H3

Inchi Key

OTVKPSUMDAFRQC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid or lump



Still not finding the right product?

Explore all of our products under 2-Bromo-4,5-dimethylpyridine



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library