Skip to Content
Merck

BDPH9BCDF4BE

Bide Pharmatech

2-Amino-1-(pyrrolidin-1-yl)ethanone

Synonym(s):

2-oxo-2-(1-pyrrolidinyl)ethanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H12O1N2
CAS Number:
Molecular Weight:
128.17
MDL number:
Product BDPH9BCDF4BE is not currently sold in your country.


Inchi Code

1S/C6H12N2O/c7-5-6(9)8-3-1-2-4-8/h1-5,7H2

Inchi Key

SJNAQBNIUKINQH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library