Skip to Content
Merck

BDPH9BCDF63D

Bide Pharmatech

5-Hydroxy-2-methoxybenzenesulfonyl chloride

Synonym(s):

5-hydroxy-2-methoxybenzenesulfonyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7O4Cl1S1
CAS Number:
Molecular Weight:
222.65
MDL number:
Product BDPH9BCDF63D is not currently sold in your country.


Inchi Code

1S/C7H7ClO4S/c1-12-6-3-2-5(9)4-7(6)13(8,10)11/h2-4,9H,1H3

Inchi Key

QPWPLOMELUAVLX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library