Skip to Content
Merck

BDPH9BCDFA3A

Bide Pharmatech

Ethyl 3-(4-((tert-butoxycarbonyl)amino)phenyl)-3-oxopropanoate

Synonym(s):

ethyl 3-{4-[(tert-butoxycarbonyl)amino]phenyl}-3-oxopropanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H21O5N1
CAS Number:
Molecular Weight:
307.35
MDL number:
Product BDPH9BCDFA3A is not currently sold in your country.


Inchi Code

1S/C16H21NO5/c1-5-21-14(19)10-13(18)11-6-8-12(9-7-11)17-15(20)22-16(2,3)4/h6-9H,5,10H2,1-4H3,(H,17,20)

Inchi Key

GRNQQSYBDFHTSO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library