Product BDPH9BCDFDE4 is not currently sold in your country.
Inchi Code
1S/C5H2BrClFN/c6-3-2-9-5(7)1-4(3)8/h1-2H
Inchi Key
IHOUCYWEAFGESV-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Country of Origin
CN
Physical Form
Solid or Semi-Solid or Liquid or Lump
Storage temp.
Inert atmosphere, 2-8°C
Still not finding the right product?
Explore all of our products under 5-Bromo-2-chloro-4-fluoropyridine
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
