Skip to Content

BDPH9BCDFE36

Bide Pharmatech

6-Azaspiro[3.4]octane-2,5-dione

Synonym(s):

6-azaspiro[3.4]octane-2,5-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H9O2N1
CAS Number:
Molecular Weight:
139.15
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H9NO2/c9-5-3-7(4-5)1-2-8-6(7)10/h1-4H2,(H,8,10)

Inchi Key

UZZSKGIHKCJBDB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Sealed in dry, 2-8C



Still not finding the right product?

Explore all of our products under 6-Azaspiro[3.4]octane-2,5-dione



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library