Skip to Content

BDPH9BCDFEDC

Bide Pharmatech

1-(5-Bromothiophen-2-yl)butan-1-one

Synonym(s):

1-(5-bromo-2-thienyl)-1-butanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9O1Br1S1
CAS Number:
Molecular Weight:
233.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H9BrOS/c1-2-3-6(10)7-4-5-8(9)11-7/h4-5H,2-3H2,1H3

Inchi Key

DUGRGUURUDYJSJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library