Skip to Content
Merck

BDPH9BCDFEEE

Bide Pharmatech

1H-indole-6-sulfonyl chloride

Synonym(s):

1H-indole-6-sulfonyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H6O2N1Cl1S1
CAS Number:
Molecular Weight:
215.66
MDL number:
Product BDPH9BCDFEEE is not currently sold in your country.


Inchi Code

1S/C8H6ClNO2S/c9-13(11,12)7-2-1-6-3-4-10-8(6)5-7/h1-5,10H

Inchi Key

XAVWTRDQHMLGOL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 1H-indole-6-sulfonyl chloride



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library