Skip to Content
Merck

BDPH9BCE0134

Bide Pharmatech

(3,5-Diamino-4-isobutylphenyl)boronic acid

Synonym(s):

(3,5-diamino-4-isobutylphenyl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H17O2N2B1
Molecular Weight:
208.07
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H17BN2O2/c1-6(2)3-8-9(12)4-7(11(14)15)5-10(8)13/h4-6,14-15H,3,12-13H2,1-2H3

Inchi Key

OUEVJUPJROEIHF-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library