Inchi Code
1S/C8H11NO3S/c9-13(11,12)8-3-1-7(2-4-8)5-6-10/h1-4,10H,5-6H2,(H2,9,11,12)
Inchi Key
RYKHXKKLCRPAJP-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 4-(2-Hydroxyethyl)benzene-1-sulfonamide
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
