Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C8H10FN/c1-6-4-7(5-10)2-3-8(6)9/h2-4H,5,10H2,1H3
Inchi Key
PQXWYQZQCWWMDQ-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under (4-Fluoro-3-methylphenyl)methanamine
hstatements
Precautionary Statements
Signal Word
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
