Skip to Content
Merck

BDPH9BCE0DED

Bide Pharmatech

6-Hydroxy-8-methylquinolin-2(1H)-one

Synonym(s):

6-hydroxy-8-methyl-2(1H)-quinolinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O2N1
CAS Number:
Molecular Weight:
175.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H9NO2/c1-6-4-8(12)5-7-2-3-9(13)11-10(6)7/h2-5,12H,1H3,(H,11,13)

Inchi Key

XKIKBJPZUYXYOL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library