Product BDPH9BCE0E4E is not currently sold in your country.
Inchi Code
1S/C6H5F3N2O2/c1-13-5-10-3(6(7,8)9)2-4(12)11-5/h2H,1H3,(H,10,11,12)
Inchi Key
ILOOCRYHYUMTKL-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
White to off-white solid
Still not finding the right product?
Explore all of our products under 2-Methoxy-6-(trifluoromethyl)pyrimidin-4(3H)-one
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
