Skip to Content
Merck

BDPH9C01E640

Bide Pharmatech

S-Methyl-s-(3-methylphenyl) sulfoximine

Synonym(s):

1-methyl-3-(methylsulfonimidoyl)benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H11O1N1S1
CAS Number:
Molecular Weight:
169.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H11NOS/c1-7-4-3-5-8(6-7)11(2,9)10/h3-6H,2H2,1H3,(H2,9,10)

Inchi Key

CDJIROMHWRNVKK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library