Skip to Content
Merck

BDPH9C01E774

Bide Pharmatech

(5'-Formyl-[2,2'-bithiophen]-5-yl)boronic acid

Synonym(s):

(5'-formyl-[2,2'-bithiophen]-5-yl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H7O3S2B1
CAS Number:
Molecular Weight:
238.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H7BO3S2/c11-5-6-1-2-7(14-6)8-3-4-9(15-8)10(12)13/h1-5,12-13H

Inchi Key

RDIOTSOSMWVMSX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library