Skip to Content

BDPH9C01EB8D

Bide Pharmatech

1-Phenylphosphinan-4-one 1-oxide

Synonym(s):

1-phenylphosphinan-4-one 1-oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H13O2P/c12-10-6-8-14(13,9-7-10)11-4-2-1-3-5-11/h1-5H,6-9H2

Inchi Key

IVCQUOFBVFOENY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 1-Phenylphosphinan-4-one 1-oxide



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library