Skip to Content
Merck

BDPH9C021D6C

Bide Pharmatech

trans-4-((Dimethylamino)methyl)cyclohexanamine dihydrochloride

Synonym(s):

(1r,4r)-4-((dimethylamino)methyl)cyclohexan-1-amine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H22N2Cl2
CAS Number:
Molecular Weight:
229.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid

Inchi Code

1S/C9H20N2.2ClH/c1-11(2)7-8-3-5-9(10)6-4-8;;/h8-9H,3-7,10H2,1-2H3;2*1H/t8-,9-;;

Inchi Key

CGFHCEZSOGYGJR-JZDLBZJVSA-N



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library