Skip to Content
Merck

BL3H160BD2CE

BLD Pharmatech

(3-Methoxy-2-nitrophenyl)methanol

Synonym(s):

(3-methoxy-2-nitrophenyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9O4N1
CAS Number:
Molecular Weight:
183.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H9NO4/c1-13-7-4-2-3-6(5-10)8(7)9(11)12/h2-4,10H,5H2,1H3

Inchi Key

YYIIAPQEOXNDLW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under (3-Methoxy-2-nitrophenyl)methanol




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library