Skip to Content
Merck

BL3H97F03434

BLD Pharmatech

(6-Bromo-2-fluoropyridin-3-yl)boronic acid

Synonym(s):

6-bromo-2-fluoro-3-pyridinylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H4O2N1Br1F1B1
CAS Number:
Molecular Weight:
219.81
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H4BBrFNO2/c7-4-2-1-3(6(10)11)5(8)9-4/h1-2,10-11H

Inchi Key

YUGZMJMNGRGYRZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

White to Yellow Solid

Storage temp.

Inert atmosphere, 2-8C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library