Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C20H24O3
CAS Number:
Molecular Weight:
312.40
UNSPSC Code:
41116107
EC Number:
233-432-5
grade
certified reference material
agency
NMI Australia certified reference material
form
solid
manufacturer/tradename
NMI Australia
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
neat
storage temp.
2-8°C
InChI
1S/C20H24O3/c1-12(21)23-19-8-7-18-17-5-3-13-11-14(22)4-6-15(13)16(17)9-10-20(18,19)2/h9-11,17-19H,3-8H2,1-2H3/t17-,18+,19+,20+/m1/s1
InChI key
CMRJPMODSSEAPL-FYQPLNBISA-N
signalword
Danger
hcodes
Hazard Classifications
Carc. 2 - Repr. 1B
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
