Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C27H38O3
CAS Number:
Molecular Weight:
410.59
UNSPSC Code:
41116107
Pricing and availability is not currently available.
grade
certified reference material
agency
NMI Australia certified reference material
form
solid
manufacturer/tradename
NMI Australia
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
neat
storage temp.
2-8°C
InChI
1S/C27H38O3/c1-26-15-13-20(28)17-19(26)8-9-21-22-10-11-24(27(22,2)16-14-23(21)26)30-25(29)12-7-18-5-3-4-6-18/h13,15,17-18,21-24H,3-12,14,16H2,1-2H3/t21-,22-,23-,24-,26-,27-/m0/s1
InChI key
QPMSXPMLTYTHGM-ZLQWOROUSA-N
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Carc. 2 - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
