Skip to Content
Merck

Skip To

COMH97E337F5

Combi-Blocks, Inc.

Phosphotungstic acid hydrate

Synonym(s):

diphosphooxidane triheptacontahydrate tetracosatungsten

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Ships in 14 days - from Aldrich Partner.
€80.77
25 g
Ships in 14 days - from Aldrich Partner.
€115.38
100 g
Ships in 14 days - from Aldrich Partner.
€205.37
500 g
Ships in 14 days - from Aldrich Partner.
€641.49

About This Item

CAS Number:
Molecular Weight:
5724.32
MDL number:

Vendor SKU: QF-5827-5G

€80.77


Ships in 14 days - from Aldrich Partner.

To order products, please contact your local dealer.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/O5P2.H2O.72O.24W/c1-6(2)5-7(3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

Inchi Key

YVZWSZZGHDWWJW-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P400679690455970
description

-

description

reagent grade

description

suitable for microscopy

description

99.995% trace metals basis (Purity excludes up to 300 ppm Si)

Purity

95%

Purity

-

Purity

-

Purity

-

Country of Origin

CN

Country of Origin

-

Country of Origin

-

Country of Origin

-

Inchi Code

1S/O5P2.H2O.72O.24W/c1-6(2)5-7(3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h;1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

Inchi Code

-

Inchi Code

-

Inchi Code

-


MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QF-5827



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library