Skip to Content
Merck

Skip To

71007-M

NovaTaqTM PCR Master Mix

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41121800
NACRES:
NA.55
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

dry ice

Quality Segment

storage temp.

−20°C

General description

The NovaTaq PCR Master Mix is a ready-to-use 2X mixture of NovaTaq DNA Polymerase, ultrapure deoxynucleotides, and reaction buffer without MgCl2. The Master Mix simplifies the assembly of PCR reactions and offers advantages of time savings, consistency, and minimal risk of contamination. Simply add the NovaTaq PCR Master Mix to an equal volume containing the required amount of MgCl2, DNA template, and primers, and the reaction is ready for thermal cycling. The final diluted reaction contains 2.5 U of NovaTaq DNA Polymerase per 100 µl. Sufficient components are included for 200 standard 50-µl (or 100 × 100-µl) amplification reactions.

Disclaimer

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patents rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California, 94404, USA.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library