Skip to Content
Merck

Skip To

MXRPC0VER

Milliflex® Autospray Station Vial Holder Cover

For Protecting Reagent Vials holders on Milliflex® Autospray Station

Synonym(s):

Vial Holder Cover

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.79
UNSPSC Code:
41116500
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polyurethane

packaging

pkg of 2 units

manufacturer/tradename

Milliflex®

parameter

15-40 °C temp. limit

application(s)

industrial qc
microbiology
pharmaceutical
sterility testing

compatibility

for use with Milliflex® Autospray System

General description

Milliflex® Autospray Station Vial Holder Cover is an accessory part that is designed to close and protect the two vial holders stationed at the top of the Milliflex® Autospray station when reagent bottles are not installed. These vial holders covers ensure that vials supports stay clean and cannot be contaminated when reagent bottles are not installed on the system.
Milliflex® Autospray Station is used for rapid microbial detection in filterable samples.

Application

Milliflex® Autospray Station Vial Holder Cover is used to protect the reagent bottles support on top of the Milliflex® Autospray station. The reagents help detect and quantify microbial presence on filtered samples.

Legal Information

MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MXRPGLASSMXRP2SPRKTMXRPCLKT1
description

For Protecting Reagent Vials holders on Milliflex® Autospray Station

description

For Spraying Reagents on the Milliflex® membrane

description

Rapid detection,response and resolution of microbial contamination

description

For 20 cleanings and 2 decontaminations

compatibility

for use with Milliflex® Autospray System

compatibility

for use with Milliflex® Autospray System

compatibility

for use with Milliflex® Rapid

compatibility

Milliflex® Rapid

packaging

pkg of 2 units

packaging

pkg of 1 unit

packaging

pkg of 1 kit

packaging

pkg of 1 kit

material

polyurethane

material

polyurethane

material

polyurethane cover, 316L stainless steel turntable(s), polyurethane case, polyester key, 316L stainless steel holder (for pad), 316L stainless steel holder (for membrane)

material

-

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

manufacturer/tradename

Milliflex®

application(s)

industrial qc
microbiology
pharmaceutical
sterility testing

application(s)

industrial qc
microbiology
pharmaceutical
sterility testing

application(s)

bioburden testing
food and beverages
pharmaceutical
sterility testing
water monitoring

application(s)

bioburden testing
environmental
food and beverages
pharmaceutical
sterility testing

parameter

15-40 °C temp. limit

parameter

15-40 °C temp. limit

parameter

15-40 °C temperature

parameter

-




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters