Inchi Code
1S/C23H44O13/c1-27-23(26)3-5-29-7-9-31-11-13-33-15-17-35-19-21-36-20-18-34-16-14-32-12-10-30-8-6-28-4-2-22(24)25/h2-21H2,1H3,(H,24,25)
Inchi Key
OBQQYTZUVFACEQ-UHFFFAOYSA-N
Storage temp.
-20
Purity
96%
Country of Origin
CN
Shipping Temperature
ambient
Physical Form
Yellow
MSDS
https://precisepeg.com/cdn/shop/files/AG-2509_SDS.pdf
hstatements
Precautionary Statements
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.