Skip to Content

PG9H9BC7B16C

PrecisePEG

endo BCN-PEG3-Alcohol

Synonym(s):

((1R,8S,9s)-bicyclo[6.1.0]non-4-yn-9-yl)methyl hydrogen (2-(2-(2-(2-hydroxyethoxy)ethoxy)ethoxy)ethyl)carbonimidate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C19H31NO6/c21-8-10-24-12-14-25-13-11-23-9-7-20-19(22)26-15-18-16-5-3-1-2-4-6-17(16)18/h16-18,21H,3-15H2,(H,20,22)/t16-,17+,18-

Inchi Key

RPISEOIEEFQRPT-BCDXTJNWSA-N

Storage temp.

-20

Purity

96%

Country of Origin

CN

Shipping Temperature

ambient

Physical Form

Yellow



MSDS

https://precisepeg.com/cdn/shop/files/AG-2032_SDS.pdf

hstatements

Precautionary Statements



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library