Skip to Content
Merck

Skip To

82002BC

Assurance® GDS Adaptor Box Power Cable ANZ

Assurance GDS®

Synonym(s):

Power Cable

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.61
UNSPSC Code:
26121629
eCl@ss:
27400102
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Assurance GDS®

technique(s)

PCR: suitable

application(s)

food and beverages

compatibility

for use with BioControl GDS

storage temp.

room temp

General description

The Assurance® GDS Adaptor Box Power Cable ANZ is an essential part to be used with the GDS Adaptor Box for the GDS Rotor-Gene Q. As part of the GDS system, the Adaptor Box improves laptop connectivity and PCR run flexibility, allowing one laptop to control up to five Rotor-Genes at one time.

Application

The Assurance® GDS Adaptor Box Power Cable ANZ is an essential part used with the Assurance® GDS Adaptor Box.

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
Assurance GDS is a registered trademark of BioControl Systems, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
82003BC82004BC82005BC
description

Assurance GDS®

description

Assurance GDS®

description

Assurance GDS®

description

Assurance GDS®

compatibility

for use with BioControl GDS

compatibility

for use with BioControl GDS

compatibility

for use with BioControl GDS

compatibility

for use with BioControl GDS

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

product line

Assurance GDS®

product line

Assurance GDS®

product line

Assurance GDS®

product line

Assurance GDS®




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters