Skip to Content
Merck

PHI90004

D-Glu-(17)-Liraglutide

GLP-1 Agonist Impurity, analytical standard, Numbering in the name as per API

Synonym(s):

D-Glu-(23)-Liraglutide, H-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-D-Glu-Ala-Ala-Lys (γ-Glu-Palmitoyl)-Glu-Phe-Ile-Ala-Trp-Leu-Val-Arg-Gly-Arg-Gly-OH

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C172H264N42O52
Molecular Weight:
3752.19
UNSPSC Code:
41116107
eCl@ss:
25140904
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

API family

liraglutide, liraglutide, liraglutide, liraglutide

form

powder

technique(s)

HPLC: suitable, LC/MS: suitable, liquid chromatography (LC): suitable, mass spectrometry (MS): suitable

color

white to off-white

format

neat

storage temp.

−20°C

SMILES string

OC[C@@H](C(N[C@@H](CC1=CC=C(O)C=C1)C(N[C@@H](CC(C)C)C(N[C@@H](CC C(O)=O)C(NCC(N[C@H](CCC(O)=O)C(N[C@@H](C)C(N[C@@H](C)C(N[C@@H](CCCCNC(CC [C@@H](C(O)=O)NC(CCCCCCCCCCCCCCC)=O)=O)C(N[C@H](C(N[C@H](C(N [C@@H]([C@@H](C)CC)C(N[C@H](C(N[C@@H](CC2=CNC3=CC=CC=C23)C(N[C@@H](CC(C) C)C(N[C@@H](C(C)C)C(N[C@H](C(NCC(N[C@H](C(NCC(O)=O)=O)CCCNC( N)=N)=O)=O)CCCNC(N)=N)=O)=O)=O)=O)C)=O)=O)CC4=CC=CC=C4)=O)CCC(O)=O)=O)=O )=O)=O)=O)=O)=O)=O)=O)NC([C@H](CO)NC([C@H](C(C)C)NC([C@H](CC (O)=O)NC([C@H](CO)NC([C@H]([C@H](O)C)NC([C@H](CC5=CC=CC=C5)N C([C@H]([C@H](O)C)NC(CNC([C@H](CCC(O)=O)NC([C@H](C)NC([C@H](CC6=CNC=N6)N )=O)=O)=O)=O)=O)=O)=O)=O)=O)=O)=O

General description

D-Glu(17)-Liraglutide is a stereoisomeric impurity of Liraglutide characterised by the replacement of L-Glutamic acid at position 17 (API numbering; position 17 in GLP-1 numbering) with its D-enantiomer (D-Glu). Position 17 resides within the central α-helical domain of the peptide, and its stereoinversion may perturb helix stability and receptor contact surface. The impurity is otherwise structurally identical to Liraglutide, retaining the palmitoyl–γGlu–Lys26 fatty acid conjugate. Racemisation at Glu residues during SPPS, particularly under prolonged activation or elevated temperature, is a recognised source of this impurity type.

Application

Used as an analytical standard for the identification and semi-quantification of the D-Glu17 diastereomeric impurity in Liraglutide API by reversed-phase HPLC, chiral chromatography, and LC-MS/MS methods.


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library