Skip to Content
Merck

A8501

Adamantane

Synonym(s):

Tricyclo[3.3.1.13,7]decane

Select a Size

Change View

About This Item

EC Number:
206-001-4
UNSPSC Code:
12352103
eCl@ss:
32150313
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Hazard Codes

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library