Skip to Content
Merck

AMAB92002

Monoclonal Anti-LHX9 antibody produced in mouse

Prestige Antibodies® Powered by Atlas Antibodies, clone CL15033, purified antibody (Protein A)

Synonym(s):

NA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
CL15033, monoclonal
Application:
WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody (Protein A)

antibody product type

primary antibodies

clone

CL15033, monoclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1 μg/mL

pH range

7.2-7.4

isotype

IgG2a

Ensembl | human accession no.

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

human ... LHX9(56956)

General description

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Immunogen

Synthetic Peptide

Application

All Prestige Antibodies Powered by Atlas Antibodies aredeveloped and validated bythe[https://https://www.proteinatlas.org]HumanProteinAtlas (HPA) project and as a result, are supported by the most extensivecharacterization in the industry.

The Human Protein Atlas project can be subdivided into threeefforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodiesthat have been generated in support of the Tissue and Cancer Atlas projectshave been tested by immunohistochemistry against hundreds of normal and diseasetissues and through the recent efforts of the Human Cell Atlas project, manyhave been characterized by immunofluorescence to map the human proteome notonly at thetissue level but now at the subcellular level. These images and thecollection of this vast data set can be viewed on the Human Protein Atlas (HPA)site by clicking on the Image Gallery link. We also provide PrestigeAntibodies®[/products/protein-biology/antibodies/prestige-antibodies]protocolsand other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

100UL pack size is provided in a solution, in PBS(phosphate-buffered saline), pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Large pack size of 1ml (at 1mg/ml) is available in PBS only, purified, biocide free, carrier free and glycerol free. Order here

Preparation Note

Upon arrival, mix gently and aliquot to avoid repeated freeze-thaw cycles

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library