Skip to Content
Merck

B3166

5-Bromo-4-chloro-3-indolyl N-acetyl-β-D-galactosaminide

~95%

Synonym(s):

X-GalNAc, 5-Bromo-4-chloro-3-indolyl 2-acetamido-2-deoxy-β-D-galactopyranoside, 5-Bromo-4-chloro-3-indolyl N-acetyl-β-D-galactosaminide, X-Galactosaminide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H18BrClN2O6
CAS Number:
Molecular Weight:
449.68
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~95%

mp

≥220 °C (lit.)

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]1Oc2c[nH]c3ccc(Br)c(Cl)c23

InChI

1S/C16H18BrClN2O6/c1-6(22)20-13-15(24)14(23)10(5-21)26-16(13)25-9-4-19-8-3-2-7(17)12(18)11(8)9/h2-4,10,13-16,19,21,23-24H,5H2,1H3,(H,20,22)/t10-,13-,14+,15-,16-/m1/s1

InChI key

SUWPNTKTZYIFQT-LMXXTMHSSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library