Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Biological source:
bovine fetus
Origin:
Australia origin
Sterility:
γ-irradiated, sterile-filtered
Form:
liquid
Pricing and availability is not currently available.
biological source
bovine fetus
sterility
γ-irradiated, sterile-filtered
form
liquid
origin
Australia origin
technique(s)
cell culture | hybridoma: suitable, cell culture | mammalian: suitable
storage temp.
−20°C
General description
Animal serum is commonly used to supplement basal media formulations for the optimal growth of many cell types in vitro.
Bovine Calf Serum (BCS) is collected from calves that are less than 12 months old. The serum contains approximately double the protein of Fetal Bovine Serum (FBS). BCS will promote growth in a variety of mammalian cell lines and is often used as a cost effective substitute for FBS.
Gamma irradiation has been shown to inactivate up to six logs of many biological contaminants in serum while maintaining growth promotion potential, providing added confidence in the quality and performance of the serum. The standard gamma irradiation of Sigma-Aldrich serum includes exposure under controlled conditions to a 60Co source with a delivered dose of 25 - 35 kGy.
Bovine Calf Serum (BCS) is collected from calves that are less than 12 months old. The serum contains approximately double the protein of Fetal Bovine Serum (FBS). BCS will promote growth in a variety of mammalian cell lines and is often used as a cost effective substitute for FBS.
Gamma irradiation has been shown to inactivate up to six logs of many biological contaminants in serum while maintaining growth promotion potential, providing added confidence in the quality and performance of the serum. The standard gamma irradiation of Sigma-Aldrich serum includes exposure under controlled conditions to a 60Co source with a delivered dose of 25 - 35 kGy.
Preparation Note
To effectively preserve the integrity of animal serum, it should be stored frozen and protected from light. Multiple thaw/freeze cycles should be avoided as they will hasten the degradation of serum nutrients and can result in the formation of insoluble precipitates.
For stability and optimal performance, serum should be stored at −20 °C and used prior to the labeled expiration date
For stability and optimal performance, serum should be stored at −20 °C and used prior to the labeled expiration date
Analysis Note
Endotoxin and hemoglobin tested
Still not finding the right product?
Explore all of our products under Bovine Calf Serum
Storage Class
10 - Combustible liquids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.