Skip to Content
Merck

Skip To

BPS53029

Kinetic HDAC Assay Developer

Synonym(s):

buffers

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41105331
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

storage temp.

-65 to -96°C

General description

The Kinetic HDAC assay developer is an essential reagent for signal generation in Kinetic HDAC assays performed with any of BPS Bioscience′s HDAC fluorimetric substrates. The developer sensitizes the deacetylated substrate so that subsequent treatment with the HDAC assay developer produces a fluorophore.
A 10x concentrate (1 ml) with 10 ml dilution buffer.

Application

Study of HDAC kinetics, regulation and inhibitor sensitivity. Suitable for use with the Kinetic HDAC assay kits:

  • Kinetic HDAC1 Assay Kit
  • Kinetic HDAC2 Assay Kit
  • Kinetic HDAC3 Assay Kit
Datasheet and Protocols may be found here.

Preparation Note

Dilute 10x developer to 1x with dilution buffer before use. Add 50 µl 1x developer per 50 µl HDAC reaction (1:1, total volume 100 µl).

Other Notes

Avoid freeze/thaw cycles.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BPS50030MAK441
description

-

description

-

description

Sufficient for 400 fluorometric tests

storage temp.

-65 to -96°C

storage temp.

-65 to -96°C

storage temp.

−20°C

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

-


Pictograms

Health hazard

Signal Word

Danger

Hazard Codes

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library