Skip to Content
Merck

Skip To

BPS60321

PDE2A Assay Kit

Synonym(s):

Assay Kits, PDE2A, phosphodiesterase 2A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41121811
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

technique(s)

activity assay: suitable (Fluorescence Polarization)

UniProt accession no.

shipped in

dry ice

storage temp.

-65 to -96°C

General description

The PDE2A Assay Kit is designed for identification of PDE2A inhibitors using fluorescence polarization.The assay is based on the binding of a fluorescent nucleotide monophosphate generated by PDE2A to the binding agent. PDE2A catalyzes the hydrolysis of the phosphodiester bond in dye-labeled cyclic adenosine monophosphate (cAMP). Nanoparticle beads selectively bind the phosphate group in the nucleotide product. This increases the size of the nucleotide relative to unreacted cAMP. Since the degree of polarization of a fluorophore is inversely related to its molecular rotation, dyes attached to the slowly-rotating nucleotide-bead complexes will not have time to reorient and therefore will emit highly polarized light. Conversely, dyes attached to the rapidly-rotating cyclic monophosphates will obtain random orientations and emit light with low polarization. The key to the PDE2A Assay Kit is the specific binding agent. Using this kit, only two simple steps on a microtiter plate are required for PDE2A reactions. First, the fluorescently labeled cAMP is incubated with a sample containing PDE2A1 for 1 hour. Second, a binding agent is added to the reaction mix to produce a change in fluorescent polarization. The FP signal is measured using a fluorescent microplate reader capable of measuring fluorescence polarization.

Application

Study enzyme kinetics and screen small molecule inhibitors for drug discovery in high throughput screening (HTS) applications.
Phosphodiesterases (PDEs) play an important role in the dynamic regulation of the second messengers cAMP(cyclic adenosine monophosphate) and cGMP (cyclic guanosine monophosphate) signaling, by hydrolyzing them. The PDE superfamily is composed of 11 families, with PDE4, 7 and 8 being cAMP-specific hydrolases, and thus regulating positive and negative responses to it. Human PDE2A has multiple biological functions, in a cell-specific context. It is involved in mitochondrial morphology and removal, apoptosis and mouse liver development and hematopoiesis. Additionally, it has been linked to cancer progression, as in colorectal cancer, melanoma, glioma and hepatocellular carcinoma (HCC). PDE2 plays a role in the PDE2/cAMP/PKA (protein kinase A) axis, with increased calcium uptake leading to higher activity of PDE2, and a decrease in the levels of TFAM (mitochondrial transcriptional factor A) being phosphorylated. TFAM is closely linked to colorectal cancer (CRC), one of the mostcommon cancers. Its various functions make it a highly relevant target for the treatment of many disorders, and the use of PDE2A inhibitors will continue to open new therapeutic avenues.
Datasheet and Protocols may be found here.

Features and Benefits

The PDE2A inhibitor screening assay kit comes in a convenient 384-well format, including purified PDE2A1 enzyme, fluorescently labeled PDE2A substrate (cAMP), binding agent, and PDE assay buffer for 384 enzyme reactions.


Still not finding the right product?

Explore all of our products under PDE2A Assay Kit


Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library