Skip to Content
Merck

C1663

Complement C9 deficient serum human

for complement assays

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

human

form

solution

technique(s)

activity assay: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... C9(735)

General description

Heritable complement C9 deficiency is fairly common in east Asian populations, particularly in Korea and Japan.

Application

For determination of complement C9 activity

Analysis Note

C9 is depleted by immunoadsorption as judged by a highly sensitive hemolytic assay.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee Jung Kang et al.
Annals of clinical and laboratory science, 35(2), 144-148 (2005-06-10)
Complement 9 deficiency is the most common complement deficiency in Japan, but it is rare in western countries. Because of Korea's geographical proximity to Japan, C9 deficiency in Korea has also been assumed to be common although this has never
R Kira et al.
Human genetics, 102(6), 605-610 (1998-08-14)
Deficiency of the ninth component of human complement (C9) is the most common complement deficiency in Japan but is rare in other countries. We studied the molecular basis of C9 deficiency in four Japanese C9-deficient patients who had suffered from
W P Kolb et al.
Journal of immunology (Baltimore, Md. : 1950), 122(5), 2103-2111 (1979-05-01)
C1q, a subcomponent of the first component of complement, has been isolated from human serum in fully hemolytically active form by affinity column chromatography and gel filtration with Bio-Gel A-5M. The affinity column was prepared by covalent coupling of purified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service