Skip to Content
Merck

Skip To

CLSETAR01

Corning® Gosselin™ L Shaped Spreader

Curved End, White PS, sterile

Synonym(s):

Cell SpreaderL-Type, L Rod Spreader, Plate Spreader

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41122100
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

sterility

sterile

packaging

case of 5000

manufacturer/tradename

Corning ETAR-01

General description

Disposable pipettes designed for accurate liquid transfer in laboratory applications, offering reliable performance for sampling, dosing, and general lab use.

Application

Microbiology / Plating

Legal Information

Corning is a registered trademark of Corning, Inc.
Gosselin is a trademark of Corning, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CLSETAR05CLSETAT04CLSPC100005
description

Curved End, White PS, sterile

description

Curved End, White PS, sterile

description

Green PS, sterile

description

capacity 1 L, sterile, Snap Cap

material

polystyrene

material

polystyrene

material

polystyrene

material

polypropylene

packaging

case of 5000

packaging

case of 6000

packaging

case of 3500

packaging

case of 80

sterility

sterile

sterility

sterile

sterility

sterile

sterility

sterile

manufacturer/tradename

Corning ETAR-01

manufacturer/tradename

Corning ETAR-05

manufacturer/tradename

Corning ETAT-04

manufacturer/tradename

Corning PC1000-05


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library