Skip to Content
Merck

D192

[D-Ala2]-Deltorphin II

>97%, powder

Synonym(s):

Tyr-D-Ala-Phe-Glu-Val-Val-Gly amide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C38H54N8O10
CAS Number:
Molecular Weight:
782.88
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

>97%

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)[C@@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@@H](C)NC(=O)[C@@H](N)Cc2ccc(O)cc2)C(C)C)C(=O)NCC(N)=O

InChI

1S/C38H54N8O10/c1-20(2)31(37(55)41-19-29(40)48)46-38(56)32(21(3)4)45-35(53)27(15-16-30(49)50)43-36(54)28(18-23-9-7-6-8-10-23)44-33(51)22(5)42-34(52)26(39)17-24-11-13-25(47)14-12-24/h6-14,20-22,26-28,31-32,47H,15-19,39H2,1-5H3,(H2,40,48)(H,41,55)(H,42,52)(H,43,54)(H,44,51)(H,45,53)(H,46,56)(H,49,50)/t22-,26+,27+,28+,31+,32+/m1/s1

InChI key

NUNBRHVOPFWRRG-RCEFDBTISA-N

Biochem/physiol Actions

Selective δ2 opioid receptor agonist
Selective δ2 opioid receptor agonist.


Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library