Skip to Content

D6413

Diloxanide furoate

Synonym(s):

2,2-Dichloro-N-(4-hydroxyphenyl)-N-methylacetamide 2-furoic acid ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H11Cl2NO4
CAS Number:
Molecular Weight:
328.15
UNSPSC Code:
51101500
PubChem Substance ID:
EC Number:
223-108-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibiotic activity spectrum

parasites

SMILES string

CN(C(=O)C(Cl)Cl)c1ccc(OC(=O)c2ccco2)cc1

InChI

1S/C14H11Cl2NO4/c1-17(13(18)12(15)16)9-4-6-10(7-5-9)21-14(19)11-3-2-8-20-11/h2-8,12H,1H3

InChI key

BDYYDXJSHYEDGB-UHFFFAOYSA-N



Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library