Skip to Content
Merck

Skip To

D9570

Dothiepin hydrochloride

≥98% (HPLC)

Synonym(s):

3-Dibenzo[b,e]thepin-11(6H)-ylidine-N,N-dimethyl-1-propanamine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H21NS·HCl
CAS Number:
Molecular Weight:
331.90
UNSPSC Code:
12352200
EC Number:
212-978-8
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

solubility

DMSO: ≥26 mg/mL

storage temp.

room temp

InChI

1S/C19H21NS.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/b17-11-;

InChI key

XUPZAARQDNSRJB-CULRIWENSA-N

Biochem/physiol Actions

Dothiepin hydrochloride is a tricyclic antidepressant.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library