Skip to Content
Merck

E0507

cis-7,8-Epoxy-2-methyloctadecane

~96%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

EC Number:
249-861-6
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~96%

storage temp.

2-8°C

Biochem/physiol Actions

Pheromone for Porthetria dispar




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library