Skip to Content
Merck

Skip To

ELITE

CHOZN® Elite Cell Line

ELITE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Technique(s):
cell culture | mammalian: suitable
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

DMSO solution

technique(s)

cell culture | mammalian: suitable

application(s)

mAb

shipped in

liquid nitrogen

storage temp.

−196°C

General description

The CHOZN® Elite cell line is a subclone of our original CHOZN®ZFN-Modified CHO Cell Line (CHOGS-1VL) that grows in suspension culture with chemically defined and animal component-free media. It is a glutamine synthetase (GS) knock out rendering the cells auxotrophic for the essential amino acid L-glutamine, which enables fast and easy selection of clones.

Features and Benefits

CHOZN®Elite cell line enables high producing clones, achieving a titer of 7-13 g/L with the standard vector, resulting in cost savings. For an easy adoption, the cell line is plug-and-play with your established CLD processes, for use in both fed-batch and perfusion processes. The cell line enables highly stable clones (85% of clones maintain ≥70% of the titer over 60 generations), a consistent and predictable growth profile (from 18 to 24 x 106 cells/mL for fed-batch) and regulatory compliance (cGMP banked, adventitious agent tested with full traceability and documentation available).

Packaging

Cells are provided in vials containing 1 mL at 10 x 106 cells/mL. Cells are cGMP banked in EX-CELL® CD CHO Fusion Medium (with 6 mM L-glutamine) and 7% DMSO.

Legal Information

A license agreement must be acquired prior to use of the cell line, or any products derived from the cell line in a clinical or commercial process. CHOZN is a registered trademark of Merck KGaA, Darmstadt, Germany.
CHOZN is a registered trademark of Merck KGaA, Darmstadt, Germany
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CHOK1CHODHFRCHOGS
description

ELITE

description

Suspension-adapted in CD media

description

-

description

-

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

technique(s)

-

shipped in

liquid nitrogen

shipped in

dry ice

shipped in

dry ice

shipped in

liquid nitrogen

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

Quality Level

500

Quality Level

200

Quality Level

200

Quality Level

-

application(s)

mAb

application(s)

-

application(s)

-

application(s)

-

form

DMSO solution

form

-

form

-

form

-




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library