Skip to Content
Merck

Key Documents

Skip To

H2139

1-Hexacosanol

≥97% (capillary GC)

Synonym(s):

Ceryl alcohol, Hexacosyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)25OH
CAS Number:
Molecular Weight:
382.71
UNSPSC Code:
12352211
NACRES:
NA.25
PubChem Substance ID:
EC Number:
208-044-4
Beilstein/REAXYS Number:
1783162
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥97% (capillary GC)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCO

InChI

1S/C26H54O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27/h27H,2-26H2,1H3

InChI key

IRHTZOCLLONTOC-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters