Skip to Content
Merck

Skip To

MTCA8403B

MTC™ Bio BenchBin™ Benchtop Biohazard Bin

capacity 3 L, with spring loaded lid, pack of 1 ea

Synonym(s):

MTC™ Bio BenchBin™ Benchtop Biohazard Bin, bio waste bin, MTC Bio A8403B, disposal bin, waste bin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
24111500
NACRES:
NB.43
eCl@ss:
32150313
Manufacturer/tradename:
MTC™ Bio A8403B
Material:
red (with black Hiohard warning)
Packaging:
pack of 1 ea
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

red (with black Hiohard warning)

packaging

pack of 1 ea

manufacturer/tradename

MTC™ Bio A8403B

capacity

3 L

General description

BenchBin™ is a personal waste container that any researcher can benefit from. Smaller autoclave bags to dispose of biological waste for subsequent processing or forget the autoclave bag and use the BenchBin™ as a waste container for non-hazardous waste. The red cover is easily removed from the inner basin for easy emptying of contents. Perfect for pipette, tips, microcentrifuge tubes, and gloves, features a push to open/close lid that keeps materials covered and your bench neat.

Features and Benefits

  • Safely dispose of tips, tubes, gloves, etc.
  • Secures and covers hazardous waste
  • Easy push open/close spring lid
  • Compact footprint

Legal Information

BenchBin is a trademark of MTC Bio
MTC is a trademark of MTC Bio

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MTCA8401BMTCA8403SKMTCA8401SK
description

capacity 3 L, with spring loaded lid, pack of 1 ea

description

capacity 1.5 L

description

includes 3L BenchBin™ biohazard bin & 400 red autoclave/biohazard bags

description

includes: BenchBin™ & 400 red autoclave/biohazard bags

manufacturer/tradename

MTC™ Bio A8403B

manufacturer/tradename

MTC™ Bio A8401B

manufacturer/tradename

MTC™ Bio A8403-SK

manufacturer/tradename

MTC™ Bio A8401-SK

material

red (with black Hiohard warning)

material

red (with black Biohazard warning)

material

red (with black Biohazard warning)

material

-

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

packaging

pack of 1 ea

capacity

3 L

capacity

1.5 L

capacity

3 L

capacity

1.5 L , bin


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library