Skip to Content
Merck

MTCC2590

MTC™ Bio MacroTubes® rack

2 tier, to hold, 16 x 5 mL macrotubes, pack of 1 ea

Synonym(s):

MTC™ Bio MacroTubes® rack, tube holder, tube rack, MTC Bio C2590, tube organiser

Select a Size

Change View

About This Item

UNSPSC Code:
41122804
NACRES:
NB.12
eCl@ss:
32032201
Manufacturer/tradename:
MTC™ Bio C2590
Feature:
to hold 16 x 5 mL macrotubes, 2 tier
Material:
acrylonitrile-butadiene-styrene (ABS)
Packaging:
pack of 1 ea
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

acrylonitrile-butadiene-styrene (ABS)

feature

to hold 16 x 5 mL macrotubes, 2 tier

packaging

pack of 1 ea

manufacturer/tradename

MTC™ Bio C2590

General description

2-Tiered rack for 5mL MacroTubes®, 16 place, 1 rack.

Features and Benefits

  • 2-tiered
  • 16-place
  • Perfect for use when pipetting

Legal Information

MTC is a trademark of MTC Bio
MacroTubes is a registered trademark of MTC Bio


Still not finding the right product?

Explore all of our products under MTC™ Bio MacroTubes® rack




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library