Skip to Content
Merck

NOS1

Nitric Oxide Synthase Detection Kit, Isotopic

sufficient for 50 assays

Synonym(s):

NOS Detection Kit

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160703
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 50 assays

foreign activity

nuclease and protease, free

Application

Sigma′s Nitric Oxide Synthase Detection System is a simple and specific assay for the measurement of nitric oxide synthase (NOS) activity. Nitric oxide (NO), formed from the amino acid L-arginine via the action of the NOS enzyme, is a unique cell signaling molecule that functions as both an intracellular and an extracellular messenger. NO has been implicated in vasodilation, neurotransmission, cytotoxicity, and inflammation.1
This kit utilizes a highly specific and sensitive assay for the direct quantitative measurement of nitric oxide synthase activity. The assay measures the enzymatic conversion of 3H-arginine to 3H-citrulline and is the standard assay for measuring NOS activity in cell lysates, tissue homogenates and purified enzyme preparations. Activity is measured in the picomole range and a positive control is included.

Disclaimer

Upon receipt, store Box 1 at −70 °C, Box 2 at room temperature.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library