Skip to Content
Merck

Skip To

OGS595

PSF-RENILLA-PHOTINUS - BI-DIRECTIONAL FIREFLY RENILLA LUCIFERASE PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
Renilla luciferase
Peptide cleavage:
no cleavage
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

buffered aqueous solution

mol wt

size 6170 bp

bacteria selection

kanamycin

origin of replication

pUC (500 copies)

peptide cleavage

no cleavage

reporter gene

Renilla luciferase

shipped in

ambient

storage temp.

−20°C

General description

This plasmid has been designed so that sequences can inserted between two reporter genes (renilla and firefly luciferase) to allow the relative transcription level in both directions to be determined by measuring the reporter luminescence. Dual luciferase reporter assay kits are available from other providers that are compatible with this plasmid. The plasmid only contains a Bgl2 site between the two genes to allow insertion.

Promoter Expression Level:

Application

This plasmid contains two luciferase reporter genes (Firefly and Renilla) in opposing directions with a Bgl2 restriction site in between the two genes to allow promoters to be inserted. This allows the transcriptional level of DNA sequences and promoters to be determined in both directions by inserting the sequences into the Bgl2 restriction site. If the sequence you would like to insert contains a Bgl2 site fragments with BamHI also have the same overhangs and can be ligated into this plasmid cut with Bgl2.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters