Skip to Content
Merck

SAB1410100

Anti-DEAF1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

NUDR, SPN, ZMYND5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 59.3 kDa

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable, western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DEAF1(10522)

Immunogen

DEAF1 (NP_066288.2, 1 a.a. ~ 565 a.a) full-length human protein.

Sequence
MEDSDSAAKQLGLAEAAAVAAAAAVAAAAAAAAGGEAEEPVLSRDEDSEEDADSEAERETPRVTAVAVMAAEPGHMDMGAEALPGPDEAAAAAAFAEVTTVTVANVGAAADNVFTTSVANAASISGHVLSGRTALQIGDSLNTEKATLIVVHTDGSIVETTGLKGPAAPLTPGPQSPPTPLAPGQEKGGTKYNWDPSVYDSELPVRCRNISGTLYKNRLGSGGRGRCIKQGENWYSPTEFEAMAGRASSKDWKRSIRYAGRPLQCLIQDGILNPHAASCTCAACCDDMTLSGPVRLFVPYKRRKKENELPTTPVKKDSPKNITLLPATAATTFTVTPSGQITTSGALTFDRASTVEATAVISESPAQGDVFAGATVQEASVQPPCRASHPEPHYPGYQDSCQIAPFPEAALPTSHPKIVLTSLPALAVPPPTPTKAAPPALVNGLELSEPRSWLYLEEMVNSLLNTAQQLKTLFEQAKHASTYREAATNQAKIHADAERKEQSCVNCGREAMSECTGCHKVNYCSTFCQRKDWKDHQHICGQSAAVTVQADEVHVAESVMEKVTV

Physical form

Solution in phosphate buffered saline, pH 7.4


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library